Skip to Content
Merck
All Photos(1)

Key Documents

V7754

Sigma-Aldrich

V5 Peptide

≥97% (HPLC), lyophilized powder

Synonym(s):

V5 Tag Peptide

Sign Into View Organizational & Contract Pricing

Select a Size

4 MG
$557.00

$557.00


Check Cart for Availability


Select a Size

Change View
4 MG
$557.00

About This Item

Empirical Formula (Hill Notation):
C67H113N17O21S
CAS Number:
Molecular Weight:
1524.78
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.56

$557.00


Check Cart for Availability

Product Name

V5 Peptide, ≥97% (HPLC), lyophilized powder

Quality Level

Assay

≥97% (HPLC)

form

lyophilized powder

mol wt

calculated mol wt 1524.78 Da

storage temp.

−20°C

Amino Acid Sequence

Cys-Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr

General description

V5 Tag Peptide is a synthetic peptide that has an amino acid sequence correspondinh to amino acids 95-108 of the non-structural protein V and to the RNA polymerase a subunit (P protein), of Paramyxovirus SV5 with an N-terminal cysteine.

Application

The peptide is useful for displacement of V5-tagged fusion proteins from anti-V5 antibody in immunoassays. The successful inhibition of antibody binding of V5 peptide demonstrates binding is specific.

Other Notes

The amino acid sequence corresponds to amino acids 95-109 of non-structural protein V and to RNA polymerase α subunit (P protein), of Paramyxovirus SV5 with an N-terminal cysteine.
Amino acid sequence is N-Cys-Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-C
Lyophilized from 0.1% TFA in H2O

Preparation Note

To prepare a stock solution, dissolve in water to a final concentration of 5 mg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eden A Dejene et al.
The Journal of biological chemistry, 295(30), 10255-10270 (2020-05-28)
Poly(A)-specific ribonuclease (PARN) is a 3'-exoribonuclease that plays an important role in regulating the stability and maturation of RNAs. Recently, PARN has been found to regulate the maturation of the human telomerase RNA component (hTR), a noncoding RNA required for
Yan-ling Zhang et al.
Archives of biochemistry and biophysics, 483(1), 45-54 (2009-01-06)
Tissue specific amplification of glucocorticoid action through NADPH-dependent reduction of inactive glucocorticoid precursors by 11beta-hydroxysteroid dehydrogenase (11beta-HSD1) contributes to the development of visceral obesity, insulin resistance and Type 2 Diabetes. Hexose-6-phosphate dehydrogenase (H6PDH) is believed to supply NADPH for the
Rui Chen et al.
PloS one, 12(12), e0190241-e0190241 (2017-12-28)
Survival of cancer cells in the harsh tumor microenvironment, characterized by oxygen and glucose deprivation, requires rapid initiation of cytoprotective measures. Metabolites whose levels change during stress are ideal signaling cues, particularly if used in post-translational modifications of stress-responsive signal
Paul Yenerall et al.
Cell chemical biology, 27(1), 105-121 (2019-12-31)
RUVBL1 and RUVBL2 (collectively RUVBL1/2) are essential AAA+ ATPases that function as co-chaperones and have been implicated in cancer. Here we investigated the molecular and phenotypic role of RUVBL1/2 ATPase activity in non-small cell lung cancer (NSCLC). We find that RUVBL1/2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service