Skip to Content
Merck
All Photos(1)

Key Documents

TTR001

Sigma-Aldrich

Acid Fast TISSUE-TROL Control Slides

mouse lung tissue containing Mycobacterium gordonae

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$363.00

$363.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 EA
$363.00

About This Item

UNSPSC Code:
12352106
NACRES:
NA.47

$363.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse lung (tissue containing Mycobacterium gordonae)

application(s)

hematology
histology

storage temp.

room temp

General description

TISSUE-TROL slides are comprised of paraffin-embedded tissue sections containing known characteristics and are used for monitoring staining performance in pathology laboratories. Acid Fast TISSUE-TROL Control Slides are the histology slides with a paraffin-embedded section of mouse lung tissue containing Mycobacterium gordonae.

Application

Histology slides with paraffin embedded section of mouse lung tissue containing Mycobacterium gordonae. Suitable for use with Acid Fast Stain (Procedure No. HT80).

Components

Box contains 24 unstained slides plus one reference slide stained using Acid Fast Stain (Procedure No. HT80).

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service