TT38440SC-EP
Eppendorf twin.tec® PCR Plates 384
skirted, PCR clean, volume 40 μL, colorless wells, pack of 25
Synonym(s):
EIA/RIA plate, immunoassay plate
About This Item
polypropylene wells
PCR clean
skirted
Recommended Products
material
colorless wells
polypropylene wells
well size
384
sterility
non-sterile
feature
skirt
PCR clean
skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951020702 (NA)
technique(s)
PCR: suitable
size
384 well
volume
40 μL
working volume
40 μL
Looking for similar products? Visit Product Comparison Guide
General description
Application
- standard PCR
- real-time PCR
- high throughput PCR
- small volume sample handling
Features and Benefits
- Enhanced rim design to optimize barcodes and streamline automated work processes.
- Stackable
- Complies with recommended footprint guidelines for small business server (SBS) environments.
- Suitable for popular 384-well thermocyclers like Eppendorf Mastercycler X50 and Mastercycler nexus.
- Maximum well volume of 40 µL
- Barcoding is available upon request for skirted and semi-skirted variants.
- Designed for quantitative real-time PCR with OptiTrack® matrix to expedite sample identification and reduce pipetting errors.
- Eppendorf twin.tec® PCR plates are ‘PCR clean’ and been batch-tested.
- Free of PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service