Skip to Content
Merck
All Photos(1)

Key Documents

GE29-0513-25

HiTrap® Q HP

Cytiva 29-0513-25, pack of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$95.00

$95.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$95.00

About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

$95.00


Check Cart for Availability

ligand

quaternary amine

description

Ion Exchanger Type (value)

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 29-0513-25

availability

not available in North America

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning

1-14

working range

2-12

suitability

suitable for bioprocess medium

Related Categories

General description

HiTrap Q HP are prepacked, ready-to-use Q Sepharose High Performance strong anion exchange columns for high-resolution, small-scale protein purification.

Application

Separation of protein using a strong anion exchange medium prepacked in the HiTrap column format.

Features and Benefits

  • 34 μm bead size for high-performance, high-resolution purifications.
  • Convenient and reproducible for fast, easy, high-performance separations either alone or connected in series.
  • Designed for use with a syringe, peristaltic pump, and chromatography systems such as AKTA design.
  • Strong quaternary ammonium (Q) anion exchanger.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to perform a purification of His-tagged membrane proteins.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.

Protocols

This page shows how to condition membrane proteins for further analysis with products from Cytiva.

This page covers the use of Sepharose High Performance media for purification of proteins, peptides or oligonucleotides, when to use them, and with which systems.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service