Skip to Content
Merck
All Photos(1)

Documents

TMO374130

Nunc® Coded Cryobank Vials in black low rack

polypropylene, capacity 1.0 mL, self-standing natural, sterile, blue cap, case of 960 ea

Synonym(s):

coded cryobank vial storage, cryostorage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121806
NACRES:
NB.11

material

blue cap
polypropylene
self-standing natural

sterility

sterile

packaging

case of 960 ea

manufacturer/tradename

Nunc 374130

technique(s)

cryopreservation: suitable

capacity

1.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

Related applications: Cell Culture Plastics

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service