T6125
2,2′-Thiodiethanol
≥98%
Synonym(s):
2,2′-Thiobis(ethanol), Bis(2-hydroxyethyl) sulfide, Thiodiethylene glycol, Thiodiglycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
S(CH2CH2OH)2
CAS Number:
Molecular Weight:
122.19
Beilstein:
1236325
EC Number:
MDL number:
UNSPSC Code:
12171500
Recommended Products
Assay
≥98%
refractive index
n20/D 1.5215 (lit.)
bp
164-166 °C/20 mmHg (lit.)
mp
−16 °C (lit.)
density
1.221 g/mL at 25 °C (lit.)
SMILES string
OCCSCCO
InChI
1S/C4H10O2S/c5-1-3-7-4-2-6/h5-6H,1-4H2
InChI key
YODZTKMDCQEPHD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service