Skip to Content
Merck
All Photos(2)

Key Documents

1.00206

Supelco

Phenol

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Hydroxybenzene, Carbolic acid, Hydroxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$196.00
1 KG
$492.00

$196.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
250 G
$196.00
1 KG
$492.00

About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-632-7
NACRES:
NA.21
grade:
ACS reagent
Assay:
99.0-100.5% (bromatometric)
technique(s):
UV/Vis spectroscopy: suitable
bp:
182 °C (lit.)
vapor pressure:
0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

$196.00


In StockDetails


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

99.0-100.5% (bromatometric)

form

crystalline solid

autoignition temp.

1319 °F

potency

660 mg/kg LD50, skin (Rat)

expl. lim.

8.6 %

technique(s)

UV/Vis spectroscopy: suitable

pH

5 (20 °C, 50 g/L in H2O)

bp

182 °C (lit.)

mp

40-42 °C (lit.)

transition temp

flash point 81 °C

solubility

84 g/L

density

1.071 g/mL at 25 °C (lit.)

bulk density

620 kg/m3

storage temp.

15-25°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: PX0510; PX0510-2; PX0510-1

Analysis Note

Assay (bromatometric): 99.0 - 100.5 %
Identity (coloring 1): passes test
Identity (coloring 2): passes test
Identity (precipitate): passes test
Appearance: colourless or faintly pink or faintly yellowish crystals or crystalline masses, deliquescent
Appearance of solution (67 g/l, water): clear and not more intensely coloured than referencesolution B6
Appearance of solution (50 g/l; water): no turbidity or insoluble matter
Acidic substances: passes test
Solidification temperature (Ph Eur): ≥ 39.5 °C
Solidification temperature (ACS): ≥ 40.5 °C
Residue on evaporation (Ph Eur): ≤ 0.05 %
Residue on evaporation (ACS): ≤ 0.05 %
Water (according to Karl Fischer): ≤ 0.5 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service