Skip to Content
Merck
All Photos(1)

Key Documents

T5699

Sigma-Aldrich

m-3M3FBS

>97% (HPLC)

Synonym(s):

2,4,6-Trimethyl-N-(3-trifluoromethyl-phenyl)-benzenesulfonamide, N-(3-Trifluoromethylphenyl)-2,4,6-trimethylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$197.00

$197.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
10 MG
$197.00

About This Item

Empirical Formula (Hill Notation):
C16H16F3NO2S
CAS Number:
Molecular Weight:
343.36
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

$197.00


In StockDetails


Request a Bulk Order

Assay

>97% (HPLC)

form

powder

color

white

mp

114-114.8 °C (lit.)

solubility

DMSO: >20 mg/mL

SMILES string

Cc1cc(C)c(c(C)c1)S(=O)(=O)Nc2cccc(c2)C(F)(F)F

InChI

1S/C16H16F3NO2S/c1-10-7-11(2)15(12(3)8-10)23(21,22)20-14-6-4-5-13(9-14)16(17,18)19/h4-9,20H,1-3H3

InChI key

ZIIUUSVHCHPIQD-UHFFFAOYSA-N

Application

m-3M3FBS has been used for MAP kinase assay in 293T cells. It has also been used for intracellular calcium analysis1. Furthermore, it has been used (at a conc. of 1 μM) as a PLC activator for sensitizing TRPA1 in HEK293t cells2.

Biochem/physiol Actions

First known direct activator of phospholipase C (activates β2, β3, γ1, γ2, δ1 isoforms).

Preparation Note

m-3M3FBS is soluble in DMSO at a concentration that is greater than 20 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander B Forster et al.
Molecular pain, 5, 17-17 (2009-04-18)
Morphine and its derivatives are key drugs in pain control. Despite its well-known analgesic properties morphine at high concentrations may be proalgesic. Particularly, short-lasting painful sensations have been reported upon dermal application of morphine. To study a possible involvement of
Jieny Gröper et al.
Cells, 9(4) (2020-04-29)
Ligand-based selectivity in signal transduction (biased signaling) is an emerging field of G protein-coupled receptor (GPCR) research and might allow the development of drugs with targeted activation profiles. Human formyl peptide receptor 1 (FPR1) is a GPCR that detects potentially
Shih-Wei Wang et al.
Journal of cellular physiology, 236(5), 3979-3990 (2020-12-01)
Prostate cancer has high metastatic potential. Men with higher urinary levels of the sleep hormone melatonin are much less likely to develop advanced prostate cancer compared with men with lower levels of melatonin. Melatonin has shown anticancer activity in experimental
Kathryn P Trogden et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(5), 2841-2854 (2018-02-07)
Vimentin is a cytoskeletal intermediate filament protein that is expressed in mesenchymal cells and cancer cells during the epithelial-mesenchymal transition. The goal of this study was to identify vimentin-targeting small molecules by using the Tocriscreen library of 1120 biochemically active
Ondrej Cerny et al.
Journal of immunology (Baltimore, Md. : 1950), 198(3), 1285-1296 (2017-01-01)
The adenylate cyclase toxin-hemolysin (CyaA) plays a key role in immune evasion and virulence of the whooping cough agent Bordetella pertussis. CyaA penetrates the complement receptor 3-expressing phagocytes and ablates their bactericidal capacities by catalyzing unregulated conversion of cytosolic ATP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service