Skip to Content
Merck
All Photos(1)

Key Documents

SML3368

Sigma-Aldrich

CP-673451

≥95% (HPLC)

Synonym(s):

1-[2-[5-(2-Methoxyethoxy)-1H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine, 1-[2-[5-(2-Methoxyethoxy)benzimidazol-1-yl]quinolin-8-yl]piperidin-4-ylamine, CP 673,451, CP 673451, CP-673,451, CP673,451, CP673451

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$143.00
50 MG
$575.00

$143.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$143.00
50 MG
$575.00

About This Item

Empirical Formula (Hill Notation):
C24H27N5O2
CAS Number:
Molecular Weight:
417.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$143.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N5(CCC(CC5)N)c1c2nc(ccc2ccc1)[n]3c4c(nc3)cc(cc4)OCCOC

InChI

1S/C24H27N5O2/c1-30-13-14-31-19-6-7-21-20(15-19)26-16-29(21)23-8-5-17-3-2-4-22(24(17)27-23)28-11-9-18(25)10-12-28/h2-8,15-16,18H,9-14,25H2,1H3

InChI key

DEEOXSOLTLIWMG-UHFFFAOYSA-N

Biochem/physiol Actions

ATP-competitive, potent and selective platelet-derived growth factor receptor (PDGFR) inhibitor against tumor growths in vivo.
CP-673451 is an ATP-competitive, potent and selective platelet-derived growth factor receptor inhibitor (PDGFR1 (β)/PDGFR2 (α) IC50 = 1/10 nM with 10 μM ATP; IC50 = 252 nM/c-kit, 450 nM/VEGFR-1/2; >450-fold reduced potency toward other angiogenic receptors and non-receptor kinases). CP-673451 potently inhibits PDGF-BB-induced receptor phosphorylation (IC50 = 6.4 nM; PDGFR1 transfectant) and effectively suppress human cancer xenografts-derived tumor in mice in vivo (ED50 ≤33 mg/kg via b.i.d. or q.d. p.o.) by inhibiting PDGFR phosphorylation and angiogenesis in tumor tissues.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Gregory Roberts et al.
Cancer research, 65(3), 957-966 (2005-02-12)
CP-673,451 is a potent inhibitor of platelet-derived growth factor beta-receptor (PDGFR-beta) kinase- and PDGF-BB-stimulated autophosphorylation of PDGFR-beta in cells (IC(50) = 1 nmol/L) being more than 450-fold selective for PDGFR-beta versus other angiogenic receptors (e.g., vascular endothelial growth factor receptor
Yuling Xi et al.
OncoTargets and therapy, 7, 1215-1221 (2014-07-23)
Lung cancer is the leading cause of cancer mortality in the world. Although some advances in lung cancer therapy have been made, patient survival is still poor. The platelet-derived growth factor receptors (PDGFRs) and their ligands play critical roles in
Feng He et al.
Journal of hepatology, 72(6), 1182-1195 (2020-02-28)
Hepatomegaly can be triggered by insulin and insulin-unrelated etiologies. Insulin acts via AKT, but how other challenges cause hepatomegaly is unknown. Since many hepatomegaly-inducing toxicants and stressors activate NRF2, we examined the effect of NRF2 activation on liver size and
Shuguang Pan et al.
American journal of translational research, 12(7), 3577-3595 (2020-08-11)
Cholangiocarcinoma (CCA) is an aggressive tumour with a poor prognosis due to its late clinical presentation and the lack of effective non-surgical therapies. Previous studies have reported that platelets are implicated in tumour invasion and metastasis, while their role and
Lu Yang et al.
Cell death and differentiation, 27(7), 2066-2080 (2020-01-24)
Lack of insight into the identity of the cells that initiate metastasis hampers the development of antimetastatic therapies. Only a tiny fraction of tumor cells termed metastasis-initiating cells (MICs) are able to successfully seed metastases, causing recurrence and therapeutic resistance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service