Skip to Content
Merck
All Photos(1)

Key Documents

T1014

Sigma-Aldrich

TNPAL-Sphingomyelin

1 mg/mL in chloroform/methanol (2:1)

Synonym(s):

Trinitrophenylaminolauroylsphingomyelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H73N6O12P
CAS Number:
Molecular Weight:
873.03
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98% (TLC)

form

liquid

concentration

1 mg/mL in chloroform/methanol (2:1)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](COP(O)(=O)OCC[N](C)(C)C)NC(=O)CCCCCCCCCCCNc1c(cc(cc1N(=O)=O)N(=O)=O)N(=O)=O

InChI

1S/C41H74N6O12P/c1-5-6-7-8-9-10-11-12-13-15-18-21-24-27-39(48)36(34-59-60(56,57)58-31-30-47(2,3)4)43-40(49)28-25-22-19-16-14-17-20-23-26-29-42-41-37(45(52)53)32-35(44(50)51)33-38(41)46(54)55/h24,27,32-33,36,39,42,48H,5-23,25-26,28-31,34H2,1-4H3,(H,43,49)(H,56,57)/b27-24+

InChI key

ZAKSUDRQGDIXGH-SOYKGTTHSA-N

Application

TNPAL-Sphingomyelin was used as substrate to determine the activity of sphingomyelinase spectrophotometrically.[1][2][3]

Biochem/physiol Actions

Sphingomyelin (SM) is a sphingolipid found in the myelin sheath and cell membranes of animals. The regulation of SM content by sphingomyelinase and SM synthase 2 affect the membrane properties and signaling.[4] Excess oxysterols result in increased amount of SM and calcium ions that is the main reason for artery blockage and atherosclerosis.[5]

Substrates

Substrate for microbial and animal sphingomyelinases.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Gatt et al.
Biochimica et biophysica acta, 530(3), 503-507 (1978-09-28)
A colored derivative of sphingomyelin was synthesized and used as substrate for several sphingomyelinases. The compound is N-omega-trinitrophenyl-aminolaurylsphingosylphosphorylcholine. The rate of hydrolysis of this substrate was compared to that of bovine brain sphingomyelin, labelled with tritium in the choline moiety.
W Vollmer et al.
Molecular microbiology, 39(6), 1610-1622 (2001-03-22)
Streptococcus pneumoniae is a major human pathogen and many interactions of this bacterium with its host appear to be mediated, directly or indirectly, by components of the bacterial cell wall, specifically by the phosphorylcholine residues which serve as anchors for
Fred A Kummerow
American journal of cardiovascular disease, 3(1), 17-26 (2013-03-06)
Despite major public health efforts, coronary heart disease continues to be the leading cause of death in the United States. Oxidized lipids contribute to heart disease both by increasing deposition of calcium on the arterial wall, a major hallmark of
A Dobrzyń et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 55(2), 305-313 (2004-06-24)
The sphingomyelin signalling pathway has been shown to function in different skeletal muscle types. The aim of the present study was to examine the effect of endurance training on the functioning of the pathway in the muscles. The experiments were
Yukinori Taniguchi et al.
Biochimica et biophysica acta, 1758(2), 145-153 (2006-04-04)
To understand the role of sphingomyelinase (SMase) in the function of biological membranes, we have investigated the effect of conversion of sphingomyelin (SM) to ceramide (Cer) on the assembly of domains in giant unilamellar vesicles (GUVs). The GUVs were prepared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service