Skip to Content
Merck
All Photos(1)

Documents

MBD0046

Sigma-Aldrich

Fungal DNA standard from Rhizoctonia solani

Suitable for PCR, sequencing and NGS, 10 ng/μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.24

Quality Level

form

liquid

concentration

10 ng/μL

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Fungal DNA standard from Rhizoctonia solani is a genomic DNA microbial standard that can serve as a standard for benchmarking the performance, along the workflow of microbiomics or meta-genomics analyses. It can also be used as a tool to increase reproducibility and allow comparison of results obtained by different labs. Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses. Rhizocotonia solani is a fungus that causes disease in plants, belonging to the phylum Basidiomycota. R. solani is a widespread fungus known to cause disease in a a variety of economically important crops including rice, tomato, maize and potato. The human gut is populated by three fungal phyla, Ascomycota, Basidiomycota, and Zygomycota. Read here how to use our standards to ensure data integrity for your microbiome research.

Application

Suitable as a standard for PCR, Sanger and next generation sequencing (NGS).

Features and Benefits

  • Individual microbial standard for microbiomics and meta-genomics workflow
  • Suitable standard for PCR, Sanger sequencing and 18S NGS
  • Improves bioinformatics analyses
  • Increases reproducibility and repeatability
  • Identity and purity of the genomic Rhizoctonia solani DNA has been confirmed by 18S NGS and gel electrophoresis

Physical form

Liquid -The genomic DNA is provided at ≥10 ng/μl concentration in TE buffer pH 8.0

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service