SYNS371204
Synthware™ Glass syringe
borosilicate glass 3.3, capacity 10 mL
Synonym(s):
Synthware S371204, glass sample syringe
About This Item
Recommended Products
material
borosilicate glass 3.3
packaging
pack of 5 ea
manufacturer/tradename
Kemtech S371204
capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service