Skip to Content
Merck
All Photos(1)

Key Documents

1.09001

Supelco

Folin-Ciocalteu′s phenol reagent

Synonym(s):

Reagent for phenol and bovine serum albumine acc. to Folin and Ciocalteu

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$123.00
500 ML
$312.00

$123.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$123.00
500 ML
$312.00

About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

$123.00


Available to ship on02 April 2025Details


Request a Bulk Order

form

liquid

pH

<0.5 (20 °C in H2O)

density

1.24 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Phytochemicals, Antioxidant and Antimicrobial Potentials and LC-MS Analysis of Centaurea parviflora Desf. Extracts: This research utilizes Folin-Ciocalteu reagent to analyze the antioxidant properties in Centaurea parviflora, emphasizing its application in identifying potent natural antioxidants which is crucial in pharmaceutical, nutraceutical, and cosmetic industries (Hechaichi et al., 2023).

  • Optimization of microwave-assisted extraction of antioxidant compounds from spring onion leaves using Box-Behnken design: This study showcases the use of Folin-Ciocalteu reagent to determine the efficiency of microwave-assisted extraction methods in isolating antioxidants, pivotal for enhancing food safety and shelf life (Aquino et al., 2023).

  • Effect of malting on phenolic compounds and radical scavenging activity in grains and breakfast cereals: Folin-Ciocalteu reagent is employed here to measure total phenolic content, providing insights into how malting influences the antioxidant properties of grains, essential for food scientists and health-focused food industries (Vingrys et al., 2022).

  • Advancing quantification methods for polyphenols in brown seaweeds-applying a selective qNMR method compared with the TPC assay: This research explores the comparative effectiveness of Folin-Ciocalteu reagent against advanced qNMR methods for quantifying polyphenols, critical for accurate and efficient antioxidant analysis in marine bioproducts (Wekre et al., 2022).

  • An Endemic Plant of the Mediterranean Area: Phytochemical Characterization of Strawberry Tree (Arbutus unedo L.) Fruits Extracts at Different Ripening Stages: Utilizing Folin-Ciocalteu reagent, this study evaluates the phenolic content and antioxidant capacity of strawberry tree fruits at various stages, highlighting its importance in developing functional foods and nutraceuticals (Scarano et al., 2022).

Analysis Note

Equivalent acid: c(H⁺) = 2 mol/l (2N)
Sensitivity (to phenol): conforms
Sensitivity (to bovine serum albumin): conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is the concentration of this reagent 2M? I need to prepare a 10% solution, how can I do this? Thanks!

    1 answer
    1. The concentration is 2 molar, with respect to acid. This does not equate to a weight percent, as there are multiple components present, in various amounts. The details of the composition are proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service