Skip to Content
Merck
All Photos(1)

Key Documents

SRP8043

Sigma-Aldrich

APRIL, Soluble mouse

recombinant, expressed in HEK 293 cells, FLAG® tagged, >95% (SDS-PAGE)

Synonym(s):

ACRP30headless:A-proliferation-inducing ligand, ACRP30headless:APRIL, ACRP30headless:CD256, TNFSF10

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$630.00

$630.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$630.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$630.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

recombinant

expressed in HEK 293 cells

tag

FLAG® tagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~34 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.02 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Tnfsf13(69583)

General description

APRIL is a cytokine that belongs to the TNF superfamily and binds to TACI and BCMA. It is implicated in the regulation of tumor cell growth, is involved in monocyte/macrophage-mediated immunological processes and functions as an important survival factor for plasmablasts and bone marrow plasma cells. APRIL is a high activity construct in which two trimeric APRIL ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively stimulates proliferation B cell. A basic amino acid sequence (QKQKKQ) close to the N-terminus of APRIL is required for binding to negatively charged sulfated glycosaminoglycan side chains of proteoglycans. Proteoglycans mediate binding of APRIL to tumor cells as well as primary lymphoid cells.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

The extracellular domain of mouse APRIL (aa 88-232) is fused at the N-terminus to mouse ACRP30headless (aa 18-110) and a FLAG®-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service