Skip to Content
Merck
All Photos(1)

Key Documents

B6681

Sigma-Aldrich

BAFF, soluble human

>95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

BLys, TALL-1, THANK, zTNF4

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$1,090.00

$1,090.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$1,090.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

$1,090.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

28 kDa by SDS-PAGE (reducing)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

BAFF is used to stimulate B cells, and is a tool for the detection of receptors on target cells.

Physical form

Lyophilized from a solution containing phosphate buffered saline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BAFF binds to the tumor necrosis factor receptor-like molecule B cell maturation antigen and is important for BAFF binds to the tumor necrosis factor receptor-like molecule B cell maturation antigen and is important f.
J.S. Thompson, et al.
The Journal of Experimental Medicine, 192, 129-129 (2001)
P Schneider et al.
The Journal of experimental medicine, 189(11), 1747-1756 (1999-06-08)
Members of the tumor necrosis factor (TNF) family induce pleiotropic biological responses, including cell growth, differentiation, and even death. Here we describe a novel member of the TNF family, designated BAFF (for B cell activating factor belonging to the TNF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service