Skip to Content
Merck
All Photos(1)

Key Documents

SRP6284

Sigma-Aldrich

Irisin human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

FNDC5 (cleaved), Fibronectin Type III Domain-containing Protein 5, Fibronectin Type III Repeat-containing Protein 2 (cleaved)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

~13 kDa

packaging

pkg of 10 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FNDC5(252995)

General description

Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin is cleaved from the type I membrane protein FNDC5 (fibronectin type III domain containing 5).[1][2] FNDC5 is mainly expressed in the muscles. Low levels of the mRNA are also present in the kidney, liver and lung.[2] The gene FNDC5 is mapped to human chromosome 1p35.1.[3]

Biochem/physiol Actions

Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin) and improves systemic metabolism by increasing energy expenditure.[1][2] It is mainly responsible for the conversion of white fat into brown fat. This conversion enhances metabolic uncoupling and caloric expenditure.[1] Irisin is always associated with beneficial responses of exercise on cellular metabolism.[2] In obese and type 2 diabetic patients, low levels of circulating irisin are characteristic of the adipose tissue and muscle.[4]

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute to 0.1 mg/mL with 100μL sterile water.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endre Kristóf et al.
Experimental cell research, 377(1-2), 47-55 (2019-02-23)
Brown and beige adipocytes contribute significantly to the regulation of whole body energy expenditure and systemic metabolic homeostasis not exclusively by thermogenesis through mitochondrial uncoupling. Several studies have provided evidence in rodents that brown and beige adipocytes produce a set
Rujun Zeng et al.
Bioscience trends, 12(6), 580-586 (2019-01-25)
Irisin is a myokine secreted from the cleavage of fibronectin type III domain-containing protein 5 (FNDC5) and has an effect on bone formation. There are limited studies about the structure of irisin and its functional unit. In order to clarify
Is irisin a human exercise gene?
Timmons JA
Nature, 488, E9-10 (2012)
Elke Albrecht et al.
Molecular metabolism, 34, 124-135 (2020-03-18)
Considerable uncertainty remains regarding the veracity of measuring myokine irisin more than seven years after its original description. Unresolved issues include the nature of transcription of the irisin precursor fibronectin type III domain containing 5 (FNDC5) gene across species, the
Gianluca Vadalà et al.
Cells, 9(6) (2020-06-21)
Physical exercise favors weight loss and ameliorates articular pain and function in patients suffering from osteoarthritis. Irisin, a myokine released upon muscle contraction, has demonstrated to yield anabolic effects on different cell types. This study aimed to investigate the effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service