Skip to Content
Merck
All Photos(1)

Key Documents

SRP8039

Sigma-Aldrich

Irisin

recombinant, expressed in CHO cells, FLAG® tagged, >95% (SDS-PAGE)

Synonym(s):

CD121a, Fibronectin type III domain-containing protein 5 (cleaved), Fibronectin type III repeat-containing protein 2 (cleaved)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$806.00

$806.00


Check Cart for Availability


Select a Size

Change View
10 μG
$806.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$806.00


Check Cart for Availability

recombinant

expressed in CHO cells

tag

FLAG® tagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~26 kDa

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FNDC5(252995)

General description

Irisin is an described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin is cleaved from the type I membrane protein FNDC5 (fibronectin type III domain containing 5).[1][2] FNDC5 is mainly expressed in the muscles. Low levels of the mRNA are also present in the kidney, liver and lung.[2] The gene encoding irisin is mapped to human chromosome 1p35.1.[3]

Biochem/physiol Actions

Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin) and improves systemic metabolism by increasing energy expenditure.[1][2] It is mainly responsible for the conversion of white fat into brown fat. This conversion enhances metabolic uncoupling and caloric expenditure.[1] Irisin is always associated with beneficial responses of exercise on cellular metabolism.[2] In obese and type 2 diabetic patients, low levels of circulating irisin are characteristic of the adipose tissue and muscle.[4]

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Irisin (human, aa 16-127 /mouse, aa 29-140) is Flag-tagged at the N-terminus. Irisin has 100% identity betwen mouse, rat, monkey and human.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is irisin a human exercise gene?
Timmons JA
Nature, 488, E9-10 (2012)
Common genetic variation in the human FNDC5 locus, encoding the novel muscle-derived 'browning' factor irisin, determines insulin sensitivity.
Staiger H, et al.
PLoS ONE, 8, e61903-e61903 (2013)
FNDC5 and irisin in humans: I. Predictors of circulating concentrations in serum and plasma and II. mRNA expression and circulating concentrations in response to weight loss and exercise.
Huh JY, et al.
Metabolism, Clinical and Experimental, 61, 1725-1725 (2012)
Irisin is expressed and produced by human muscle and adipose tissue in association with obesity and insulin resistance.
Moreno-Navarrete JM, et al.
The Journal of Clinical Endocrinology and Metabolism, 98, E769-E778 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service