Skip to Content
Merck
All Photos(1)

Key Documents

SRP6113

Sigma-Aldrich

UGDH human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

GDH, UDP-GlcDH, UDP-glucose dehydrogenase, UDPGDH, UGD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

59.5 kDa (533 aa, 1-494 aa + NT His-Tag)

packaging

pkg of 50 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UGDH(7358)

General description

UDP-glucose 6-dehydrogenase (UGDH) is encoded by the gene mapped to human chromosome 4p14. UGDH is part of the UDP-glucose/GDP-mannose dehydrogenase family and is a widely expressed enzyme localized in the liver.

Biochem/physiol Actions

UDP-glucose 6-dehydrogenase (UGDH) transfers UDP-glucose to UDP-glucuronate and thus takes part in the biosynthesis of glycosaminoglycans such as hyaluronan, chondroitin sulfate, and heparan sulfate. UGDH is implicated in various cellular processes such as, detoxification, polysaccharide biosynthesis and embryonic development.

Physical form

0.5 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1 mM DTT, 0.1 M NaCl, and 1 mM EDTA.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSELEM FEIKKICCIG AGYVGGPTCS VIAHMCPEIR VTVVDVNESR INAWNSPTLP IYEPGLKEVV ESCRGKNLFF STNIDDAIKE ADLVFISVNT PTKTYGMGKG RAADLKYIEA CARRIVQNSN GYKIVTEKST VPVRAAESIR RIFDANTKPN LNLQVLSNPE FLAEGTAIKD LKNPDRVLIG GDETPEGQRA VQALCAVYEH WVPREKILTT NTWSSELSKL AANAFLAQRI SSINSISALC EATGADVEEV ATAIGMDQRI GNKFLKASVG FGGSCFQKDV LNLVYLCEAL NLPEVARYWQ QVIDMNDYQR RRFASRIIDS LFNTVTDKKI AILGFAFKKD TGDTRESSSI YISKYLMDEG AHLHIYDPKV PREQIVVDLS HPGVSEDDQV SRLVTISKDP YEACDGAHAV VICTEWDMFK ELDYERIHKK MLKPAFIFDG RRVLDGLHNE LQTIGFQIET IGKKVSSKRI PYAPSGEIPK FSLQDPPNKK PKV

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment1 of the UGDH locus encoding UDP-glucose dehydrogenase to human chromosome band 4p15. 1 by radiation hybrid mapping.
Marcu O, et al.
Cytogenetic and genome research, 86(3-4), 244-245 (1999)
UDP-glucose dehydrogenase activity and optimal downstream cellular function require dynamic reorganization at the dimer-dimer subunit interfaces.
Hyde A S, et al.
The Journal of Biological Chemistry, M113-M113 (2013)
cDNA cloning and expression analysis of the human UDPglucose dehydrogenase.
Peng H L, et al.
Proceedings of the National Science Council, Republic of China. Part B, Life Sciences, 22(4), 166-172 (1998)
Ping Cai et al.
Bioscience, biotechnology, and biochemistry, 79(7), 1090-1093 (2015-03-15)
An NADPH-dependent sorbose reductase from Candida albicans was identified to catalyze the asymmetric reduction of ethyl 4-chloro-3-oxobutanoate (COBE). The activity of the recombinant enzyme toward COBE was 6.2 U/mg. The asymmetric reduction of COBE was performed with two coexisting recombinant
Prognostic significance of genetic alterations detected by high-density single nucleotide polymorphism array in gastric cancer.
Tada M, et al.
Cancer Science, 101(5), 1261-1269 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service