Skip to Content
Merck
All Photos(1)

Key Documents

SRP5219

Sigma-Aldrich

KAT3A (518-1207), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CBP, CREBBP, KAT3A, RSTS

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$321.00

$321.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$321.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$321.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~140 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CREBBP(1387)

Biochem/physiol Actions

KAT3A has intrinsic histone acetyltransferase activity that acts as a scaffold to stabilize additional protein interactions with the transcription complex and acetylates both histone and non-histone proteins. KAT3A (CREBBP) expressed as a nuclear protein that binds to cAMP-response element binding protein (CREB) and involved in the transcriptional coactivation of many different transcription factors. KAT3A plays a main role in embryonic development, growth control, and homeostasis by coupling chromatin remodeling to transcription factor recognition. KAT3A also plays a critical role in the transmission of inductive signals from cell surface receptors to the transcriptional apparatus.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lawryn H Kasper et al.
Nature, 419(6908), 738-743 (2002-10-18)
The coactivators CBP (Cre-element binding protein (CREB)-binding protein) and its paralogue p300 are thought to supply adaptor molecule and protein acetyltransferase functions to many transcription factors that regulate gene expression. Normal development requires CBP and p300, and mutations in these
J Arias et al.
Nature, 370(6486), 226-229 (1994-07-21)
A number of signalling pathways stimulate transcription of target genes through nuclear factors whose activities are primarily regulated by phosphorylation. Cyclic AMP regulates the expression of numerous genes, for example, through the protein kinase-A (PKA)-mediated phosphorylation of transcription factor CREB

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service