Skip to Content
Merck
All Photos(2)

Key Documents

SRP5209

Sigma-Aldrich

BMPR2 (174-end), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BMPR-II, BMPR3, BMR2, BRK-3, FLJ41585, FLJ76945, PPH1, T-ALK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$655.00

$655.00


Check Cart for Availability


Select a Size

Change View
10 μG
$655.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$655.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

11.9-16.1 nmol/min·mg

mol wt

~115 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BMPR2(659)

General description

BMPR2 is a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs that are involved in endochondral bone formation and embryogenesis. The loss of interaction and lack of phosphorylation of TCTEL1 by BMPR2 may contribute to the pathogenesis of primary pulmonary hypertension (PPH) and BMPR2 also plays an essential role in human T-cell differentiation.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajiv D Machado et al.
Human molecular genetics, 12(24), 3277-3286 (2003-10-30)
Diverse heterozygous mutations of bone morphogenetic receptor type II (BMPR-II) underlie the inherited form of the vascular disorder primary pulmonary hypertension (PPH). As yet, the molecular detail of how such defects contribute to the pathogenesis of PPH remains unclear. BMPR-II
Teresa Cejalvo et al.
Immunology, 121(1), 94-104 (2007-04-12)
T-cell differentiation is driven by a complex network of signals mainly derived from the thymic epithelium. In this study we demonstrate in the human thymus that cortical epithelial cells produce bone morphogenetic protein 2 (BMP2) and BMP4 and that both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service