Skip to Content
Merck
All Photos(1)

Key Documents

SML2851

Sigma-Aldrich

Galunisertib

≥98% (HPLC)

Synonym(s):

2-(6-Methyl-pyridin-2-yl)-3-[6-amido-quinolin-4-yl)-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole, 4-(2-(6-Methylpyridin-2-yl)-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazol-3-yl)quinoline-6-carboxamide, 4-[5,6-Dihydro-2-(6-methyl-2-pyridinyl)-4H-pyrrolo[1,2-b]pyrazol-3-yl]-6-quinolinecarboxamide, LY 2157299, LY-2157299, LY2157299

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$169.00
25 MG
$593.00

$169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$169.00
25 MG
$593.00

About This Item

Empirical Formula (Hill Notation):
C22H19N5O
CAS Number:
Molecular Weight:
369.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$169.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]21nc(c(c2CCC1)c4c5c(ncc4)ccc(c5)C(=O)N)c3nc(ccc3)C

InChI

1S/C22H19N5O/c1-13-4-2-5-18(25-13)21-20(19-6-3-11-27(19)26-21)15-9-10-24-17-8-7-14(22(23)28)12-16(15)17/h2,4-5,7-10,12H,3,6,11H2,1H3,(H2,23,28)

InChI key

IVRXNBXKWIJUQB-UHFFFAOYSA-N

Biochem/physiol Actions

Galunisertib (LY2157299) is an orally active multikinase inhibitor (IC50 in nM = 80/ALK4, 170/TGFβR1, 210/TGβ?R2, 190/MINK (MAP4K6), 220/RIP2 (RIPK2), 260/CK1α, 280/MEKKK4, 310/GAK, 400/CK1ε, 470/ALK6 (BMPR1B), 500/B-raf, 510/TNIK) mostly cited for its ALK4 & TGFbeta (TGFβ) receptors (TGFβ-RI (ALK5) & TGFβ-RII) inhibitory activity and anticancer efficacy in vitro (0.1-10 μM) and in vivo (20-100 mg/kg p.o.; mice).
Orally active multikinase inhibitor best known for its ALK4, TGFbeta receptors TGFβ-RI (ALK5) & TGFβ-RII activity and anticancer efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linlin Shi et al.
Theranostics, 9(14), 4115-4129 (2019-07-10)
Galunisertib (Gal) is a transforming growth factor (TGF-β) blockade which is being investigated as a potential tumor immunotherapy candidate drug in clinical trials. However, primary or acquired resistance is often found in the recruited cancer patients, which limits its clinical
Stephan Herbertz et al.
Drug design, development and therapy, 9, 4479-4499 (2015-08-27)
Transforming growth factor-beta (TGF-β) signaling regulates a wide range of biological processes. TGF-β plays an important role in tumorigenesis and contributes to the hallmarks of cancer, including tumor proliferation, invasion and metastasis, inflammation, angiogenesis, and escape of immune surveillance. There
Minsuk Kwon et al.
Immune network, 18(6), e45-e45 (2019-01-09)
Immune checkpoint inhibitors (ICIs), such as anti-PD-1 and anti-PD-L1 Abs, have shown efficacy for the treatment of various cancers. Although research has actively sought to develop new ICIs and immunomodulators, no efficient in vitro assay system is available to evaluate
Wenlong Li et al.
International journal of cancer, 146(6), 1631-1642 (2019-07-16)
Galunisertib (LY2157299), a promising small-molecule inhibitor of the transforming growth factor-beta (TGF-β) receptor, is currently in mono- and combination therapy trials for various cancers including glioblastoma, hepatocellular carcinoma and breast cancer. Using genetically modified mouse models, we investigated the roles
Mariafrancesca Cascione et al.
International journal of molecular sciences, 20(1) (2019-01-02)
The epithelial mesenchymal transition (EMT) is a physiological multistep process involving epithelial cells acquiring a mesenchymal-like phenotype. It is widely demonstrated that EMT is linked to tumor progression and metastasis. The transforming growth factor (TGF)-β pathways have been widely investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service