Skip to Content
Merck
All Photos(1)

Key Documents

SRP5160

Sigma-Aldrich

14-3-3 σ, untagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3 σ, SFN, YWHAS, stratifin

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$275.00

$275.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
$275.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$275.00


Available to ship on17 April 2025Details


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~29 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SFN(2810)

General description

14-3-3σ or stratifin is a protein that is strongly induced by gamma irradiation and other DNA-damaging agents. The induction of 14-3-3σ is mediated by a p53 responsive element. Exogenous introduction of 14-3-3σ into cycling cells results in a G2 cell cycle arrest. Knockout of 14-3-3σ in cells showed that the cells are unable to maintain cell cycle arrest after DNA damage. The 14-3-3σ -/- cells die as they enter mitosis. This process is associated with a failure of the 14-3-3σ-deficient cells to sequester the proteins that initiate mitosis and prevent them from entering the nucleus. Thus, 14-3-3σ plays an important role in maintaining the G2 checkpoint in cells and preventing mitotic death.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Chan et al.
Nature, 401(6753), 616-620 (1999-10-19)
14-3-3Sigma is a member of a family of proteins that regulate cellular activity by binding and sequestering phosphorylated proteins. It has been suggested that 14-3-3sigma promotes pre-mitotic cell-cycle arrest following DNA damage, and that its expression can be controlled by
H Hermeking et al.
Molecular cell, 1(1), 3-11 (1998-07-11)
Exposure of colorectal cancer (CRC) cells to ionizing radiation results in a cell-cycle arrest in G1 and G2. The G1 arrest is due to p53-mediated induction of the cyclin-dependent kinase inhibitor p21WAF1/CIP1/SDI1, but the basis for the G2 arrest is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service