Skip to Content
Merck
All Photos(1)

Key Documents

SML2838

Sigma-Aldrich

AP20187

≥98% (HPLC), powder, protein dimerization inducer

Synonym(s):

(2S,2′S)-1-[(2S)-1-oxo-2-(3,4,5-trimethoxyphenyl)butyl]-2-piperidinecarboxylic acid, 2,2′-[[2-[(dimethylamino)methyl]-1,3-propanediyl]bis[imino(2-oxo-2,1-ethanediyl)oxy-3,1-phenylene[(1R)-3-(3,4-dimethoxyphenyl)propylidene]]] ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$230.00
5 MG
$925.00

$230.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$230.00
5 MG
$925.00

About This Item

Empirical Formula (Hill Notation):
C82H107N5O20
CAS Number:
Molecular Weight:
1482.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$230.00


Check Cart for Availability

Request a Bulk Order

Product Name

AP20187, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N2([C@@H](CCCC2)C(=O)O[C@H](CCc8cc(c(cc8)OC)OC)c3cc(ccc3)OCC(=O)NCC(CN(C)C)CNC(=O)COc4cc(ccc4)[C@H](OC(=O)[C@H]6N(CCCC6)C(=O)[C@@H](CC)c7cc(c(c(c7)OC)OC)OC)CCc5cc(c(cc5)OC)OC)C(=O)[C@@H](CC)c1cc(c(c(c1)OC)OC)OC

InChI

1S/C82H107N5O20/c1-15-61(57-43-71(98-9)77(102-13)72(44-57)99-10)79(90)86-37-19-17-27-63(86)81(92)106-65(33-29-52-31-35-67(94-5)69(39-52)96-7)55-23-21-25-59(41-55)104-50-75(88)83-47-54(49-85(3)4)48-84-76(89)51-105-60-26-22-24-56(42-60)66(34-30-53-32-36-68(95-6)70(40-53)97-8)107-82(93)64-28-18-20-38-87(64)80(91)62(16-2)58-45-73(100-11)78(103-14)74(46-58)101-12/h21-26,31-32,35-36,39-46,54,61-66H,15-20,27-30,33-34,37-38,47-51H2,1-14H3,(H,83,88)(H,84,89)/t61-,62-,63-,64-,65+,66+/m0/s1

InChI key

NSBGUMKAXUXKGI-BPNHAYRBSA-N

Biochem/physiol Actions

AP20187 is a synthetic and cell-permeable FK506 (tacrolimus) analog that acts as a chemical dimerizer, a chemical inducer of dimerization (CID), to reversibly dimerize and activate fusion proteins containing a growth factor receptor signaling DmrB domain. It has been used to induce oligomerization of cell surface receptor proteins and to study the activity and/or re-localization of any protein that affected by oligomerization. AP20187 has been found to act as a senolytic. In a transgenic mouse model that expresses the FK506-binding protein–caspase 8 (FKBP–Casp8) fusion protein, AP20187 activated FKBP-fused Casp8 in senescent cells resulting in apoptosis of the cells resulting in delayed tumorigenesis , diminished age-related organ deterioration, and extended life span.
Synthetic and cell-permeable FK506 (tacrolimus) analog that acts as a dimerizer and activator of fusion proteins; senolytic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David W Chang et al.
The Journal of biological chemistry, 278(19), 16466-16469 (2003-03-15)
Proteolytic activation of initiator procaspases is a crucial step in the cellular commitment to apoptosis. Alternative models have been postulated for the activation mechanism, namely the oligomerization or induced proximity model and the allosteric regulation model. While the former holds
Darren J Baker et al.
Nature, 530(7589), 184-189 (2016-02-04)
Cellular senescence, a stress-induced irreversible growth arrest often characterized by expression of p16(Ink4a) (encoded by the Ink4a/Arf locus, also known as Cdkn2a) and a distinctive secretory phenotype, prevents the proliferation of preneoplastic cells and has beneficial roles in tissue remodelling
Aurelie Bedel et al.
Stem cells translational medicine, 6(2), 382-393 (2017-02-14)
Iatrogenic tumorigenesis is a major limitation for the use of human induced pluripotent stem cells (hiPSCs) in hematology. The teratoma risk comes from the persistence of hiPSCs in differentiated cell populations. Our goal was to evaluate the best system to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service