Skip to Content
Merck
All Photos(2)

Key Documents

SRP5075

Sigma-Aldrich

PTPN2, active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PTPT, TCELLPTP, TCPTP

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$449.00

$449.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$449.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$449.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

6928-9372 nmol/min·mg

mol wt

~69 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN2(5771)

Related Categories

General description

Research Area: Cell Signaling
Protein tyrosine phosphatase, non-receptor type 2 (PTPN2), is one of the most abundant mammalian tyrosine phosphatase. The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. By virtue of protein tyrosine phosphatase activity, PTPN2 is known to be a signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation involved in cell communication and signal transduction.

Biochem/physiol Actions

Protein tyrosine phosphatase, non-receptor type 2 (PTPN2) regulates interferon signaling and the endoplasmic reticulum stress response in pancreatic β-cells during autoimmune diabetes.[1] It serves as a crucial prognostic marker that influences the progression of pancreatic adenocarcinoma (PAAD) by activating the JAK-STAT signaling pathway. Silencing PTPN2 leads to a significant reduction in PAAD cell growth, migration, and invasion, while also triggering cell cycle arrest and apoptosis in PAAD cells.[2]

Physical form

Supplied in 20mM MOPS, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25mM DTT, 0.1mM PMSF, 30% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTPN2, A key predictor of prognosis for pancreatic adenocarcinoma, significantly regulates cell cycles, apoptosis, and metastasis
Kuang W, et al.
Frontiers in Immunology, 13, 805311-805311 (2022)
PTPN2 regulates the interferon signaling and endoplasmic reticulum stress response in pancreatic beta cells in autoimmune diabetes
Elvira B, et al.
Diabetes, 71(4), 653-668 (2022)
Tetsuya Yamamoto et al.
Biochemical and biophysical research communications, 297(4), 811-817 (2002-10-03)
In the previous study, we demonstrated that the nuclear isoform of T-cell protein-tyrosine phosphatase (TC-PTP) dephosphorylated and deactivated signal transducer and activator of transcription 5a (STAT5a) and STAT5b, thereby negatively regulating prolactin (PRL)-mediated signaling pathway. In this study, we examined
Sanjeev Gupta et al.
FEBS letters, 532(1-2), 61-66 (2002-12-03)
PTP-S2/TC45 is a nuclear protein tyrosine phosphatase, which induces p53-dependent apoptosis. Here we show that the p53 protein level increased in MCF-7 cells in response to PTP-S2 overexpression. PTP-S2-induced p53 protein was transcriptionally active and it could activate caspase-1 gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service