Skip to Content
Merck
All Photos(1)

Key Documents

SML2525

Sigma-Aldrich

Tafluprost

≥98% (HPLC)

Synonym(s):

(5Z)-7-[(1R,2R,3R,5S)-2-[(1E)-3,3-Difluoro-4-phenoxy-1-buten-1-yl]-3,5-dihydroxycyclopentyl]-5-heptenoic acid 1-methylethyl ester, 16-phenoxy-15-deoxy-15,15-difluoro-17,18,19,20-tetranorprostaglandin F2α isopropyl ester, AFP-168

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$287.00
25 MG
$1,090.00

$287.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$287.00
25 MG
$1,090.00

About This Item

Empirical Formula (Hill Notation):
C25H34F2O5
CAS Number:
Molecular Weight:
452.53
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$287.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

oil

optical activity

[α]/D +20 to +25°, c = 1 in chloroform-d

color

colorless to yellow

storage temp.

−20°C

SMILES string

FC(F)(COc2ccccc2)\C=C\C1C(CC(C1C\C=C\CCCC(=O)OC(C)C)O)O

InChI

1S/C25H34F2O5/c1-18(2)32-24(30)13-9-4-3-8-12-20-21(23(29)16-22(20)28)14-15-25(26,27)17-31-19-10-6-5-7-11-19/h3,5-8,10-11,14-15,18,20-23,28-29H,4,9,12-13,16-17H2,1-2H3/b8-3+,15-14+

InChI key

WSNODXPBBALQOF-ZHDUIJGQSA-N

Biochem/physiol Actions

Prostaglandin F2α agonist; Antiglaucoma agent
Taflupost is a prostaglandin F2α agonist that lowers intraocular pressure and is used in the treatment of glaucoma. It has high affinity and selectivity for the fluoroprostaglandin (FP) receptor PGF2α. Tafluprost is believed to reduce the intraocular pressure (IOP) by increasing the outflow of aqueous humor. Tafluprost is a pro-drug. The active metabolite is the free acid.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service