Skip to Content
Merck
All Photos(2)

Key Documents

SRP0721

Sigma-Aldrich

AKT1 (E17K), Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$764.00

$764.00


Check Cart for Availability


Select a Size

Change View
10 μG
$764.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$764.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

50-75 nmol/min·mg (radiometric)

mol wt

~85 kDa

concentration

0.1 μg/μL

accession no.

NM_005163

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AKT1(207)

Application

Useful for kinase assay.

Biochem/physiol Actions

Recombinant full-length human AKT1 (E17K) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT1/PKBalpha is a serine/threonine kinase that belongs to the AKT family. AKT1 is activated in cells in response to diverse stimuli such as hormones, growth factors and extracellular matrix components and is involved in glucose metabolism, transcription, survival, cell proliferation, angiogenesis, and cell motility (1). AKT1 is frequently overexpressed and active in many types of human cancers including cancers of colon, breast, brain, pancreas and prostate as well as lymphomas and leukemias (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service