Skip to Content
Merck
All Photos(1)

Key Documents

SRP0488

Sigma-Aldrich

PLA2G12A human

recombinant, expressed in mammalian cells, ≥80% (SDS-PAGE)

Synonym(s):

GXII, Phospholipase A2 G12, ROSSY

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$986.00

$986.00


Estimated to ship on30 April 2025



Select a Size

Change View
20 μG
$986.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$986.00


Estimated to ship on30 April 2025


biological source

human

recombinant

expressed in mammalian cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

22 kDa

packaging

pkg of 10 and 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.14 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

Human PLA2G12A (GenBank Accession No. NM_030821), amino acids 23-189(end) with C-terminal His-tag, MW=22 kDa, expressed in a mammalian cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Fomulated in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 200 mM Imidazole, and 20% Glycerol.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianne Berg et al.
Molecular cancer, 9, 100-100 (2010-05-13)
Estimates suggest that up to 30% of colorectal cancers (CRC) may develop due to an increased genetic risk. The mean age at diagnosis for CRC is about 70 years. Time of disease onset 20 years younger than the mean age

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service