Skip to Content
Merck
All Photos(1)

Key Documents

SRP0348

Sigma-Aldrich

Flt1 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

VEGFR1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$760.00

$760.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$760.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$760.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

67.5 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FLT1(2321)

General description

Human Flt1 ( GenBank Accession No. AF063657), amino acids 783-end with N-terminal His-tag, MW=67.5 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will transfer 1 nmol phosphate to the tyrosine substrate per minute at pH 7.4 and 30°C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Shibuya
The international journal of biochemistry & cell biology, 33(4), 409-420 (2001-04-20)
Vascular endothelial growth factor receptor-1 (VEGFR-1/Flt-1) is structurally a typical tyrosine kinase receptor of about 180 kDa, and carries seven Ig-like domains in the extracellular region and a tyrosine kinase domain with a long kinase insert. Recent studies have revealed
N Rahimi et al.
The Journal of biological chemistry, 275(22), 16986-16992 (2000-04-05)
Vascular endothelial growth factor (VEGF) provokes angiogenesis in vivo and stimulates growth and differentiation of endothelial cells in vitro. Although VEGF receptor-1 (VEGFR-1) and VEGFR-2 are known to be high affinity receptors for VEGF, it is not clear which of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service