Skip to Content
Merck
All Photos(1)

Key Documents

F6307

Sigma-Aldrich

FLT1 (784-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

VEGFR1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$609.00

$609.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$609.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$609.00


Estimated to ship on27 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

30-40 nmol/min·mg

mol wt

~94 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FLT1(2321)

General description

FLT1 (fms-related tyrosine kinase 1) or VEGFR (vascular endothelial growth factor receptor) is one of the receptor tyrosine kinase, which acts as a receptor for VEGF. VEGF is a predominant angiogenesis stimulating factor. FLT1 is a transmembrane receptor.[1] Soluble FLT1 (sFLT1) is an alternatively spliced soluble form of FLT1, expressed by multiple tissues. It acts as a negative regulator of VEGF. It does not contain the cytosolic and transmembrane domains.[2]

Biochem/physiol Actions

FLT1 (fms-related tyrosine kinase 1) or VEGFR (vascular endothelial growth factor receptor) is implicated in angiogenesis in tumors, which plays a key role in tumorigenesis, invasion and metastasis of tumors, especially the malignant ones.[1] In human circulation, angiogenesis is balanced by soluble FLT1 (sFLT1) and VEGF. An imbalance leads to multiple conditions such as pre-eclampsia.[2] sFLT1 plays an essential role in the angiogenesis of placenta. An overexpression of this protein in placenta is associated with prenatal arsenic exposure and thus, reduced birth weight.[3]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Ling Zhai et al.
PloS one, 9(7), e101779-e101779 (2014-07-10)
Endothelial injury, which may present clinically as hypertension, proteinuria and increased von Willebrand Factor (vWF) level, is a common manifestation in IgA nephropathy (IgAN). However, causal factors for endothelial injury in IgAN are not completely understood. An imbalance of vascular
Sylvie Remy et al.
PloS one, 9(3), e92677-e92677 (2014-03-26)
There is increasing epidemiologic evidence that arsenic exposure in utero is associated with adverse pregnancy outcomes and may contribute to long-term health effects. These effects may occur at low environmental exposures but the underlying molecular mechanism is not clear. We
Monica Autiero et al.
Nature medicine, 9(7), 936-943 (2003-06-11)
Therapeutic angiogenesis is likely to require the administration of factors that complement each other. Activation of the receptor tyrosine kinase (RTK) Flk1 by vascular endothelial growth factor (VEGF) is crucial, but molecular interactions of other factors with VEGF and Flk1
Hui Wang et al.
Mutagenesis, 29(5), 311-317 (2014-06-04)
Lung inflammation and epithelial to mesenchymal transition (EMT) are two pathogenic features for the two contextual diseases: chronic obstructive pulmonary disease (COPD) and lung cancer. VEGFR1 (or FLT1) plays a certain role in promoting tumour growth, inflammation and EMT. To
G H Fong et al.
Nature, 376(6535), 66-70 (1995-07-06)
The vascular endothelial growth factor (VEGF) and its high-affinity binding receptors, the tyrosine kinases Flt-1 and Flk-1, are thought to be important for the development of embryonic vasculature. Here we report that Flt-1 is essential for the organization of embryonic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service