Skip to Content
Merck
All Photos(1)

Key Documents

SML3812

Sigma-Aldrich

Asunaprevir

≥98% (HPLC)

Synonym(s):

Asunaprevir, BMS650032, 3-Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}-L-valyl-(4R)-4-[(7-chloro-4-methoxy-1-isoquinolinyl)oxy]-N-{(1R,2S)-1-[(cyclopropylsulfonyl)carbamoyl]-2-vinylcyclopropyl}-L-prolinamide, ASV, BMS 650032, BMS-650032

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$327.00
50 MG
$1,310.00

$327.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$327.00
50 MG
$1,310.00

About This Item

Empirical Formula (Hill Notation):
C35H46ClN5O9S
CAS Number:
Molecular Weight:
748.29
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$327.00


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC(C)(C)[C@@H](C(=O)N1C[C@@H](C[C@H]1C(=O)N[C@@]2(C[C@H]2C=C)C(=O)NS(=O)(=O)C3CC3)OC4=NC=C(C5=C4C=C(C=C5)Cl)OC)NC(=O)OC(C)(C)C

InChI

1S/C35H46ClN5O9S/c1-9-19-16-35(19,31(44)40-51(46,47)22-11-12-22)39-28(42)25-15-21(49-29-24-14-20(36)10-13-23(24)26(48-8)17-37-29)18-41(25)30(43)27(33(2,3)4)38-32(45)50-34(5,6)7/h9-10,13-14,17,19,21-22,25,27H,1,11-12,15-16,18H2,2-8H3,(H,38,45)(H

InChI key

XRWSZZJLZRKHHD-QGQNBIQGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Asunaprevir (ASV; BMS-650032) is an orally active, potent and selective hepatitis C virus (HCV) NS3 protease inhibitor (HCV NS3/4A IC50 = 0.7 nM; GBV-B NS3/4A IC50 = 21 µM, IC50 >5 µM using 11 human serine and cysteine proteases) that targets the NS3/4A protease complex with high affinity (Ki = 0.4 and 0.24 nM, using genotypes 1a (H77) and 1b (J4L6S), respectively). ASV inhibits HCV replication in cultures (IC50 = 1-4 nM/replicon genotypes 1 & 4, 67 nM-1.162 µM/replicon genotypes 2 & 3, >12 μM against a panel of other RNA viruses).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combination treatment with hepatitis C virus protease and NS5A inhibitors is effective against recombinant genotype 1a, 2a, and 3a viruses
Antimicrobial Agents and Chemotherapy, 57(3), 1291-1303 (2013)
Combinations of lambda interferon with direct-acting antiviral agents are highly efficient in suppressing hepatitis C virus replication
Antimicrobial Agents and Chemotherapy, 57(3), 1312-1322 (2013)
Preclinical Profile and Characterization of the Hepatitis C Virus NS3 Protease Inhibitor Asunaprevir (BMS-650032)
Antimicrobial Agents and Chemotherapy, 56(10), 5387-5396 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service