Skip to Content
Merck
All Photos(1)

Key Documents

SML3617

Sigma-Aldrich

NBP

≥95% (HPLC)

Synonym(s):

(S)-2-(6-(((p-tolylthio)carbonyl)oxy)benzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid, NE bioluminescent probe

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$163.00
25 MG
$657.00

$163.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$163.00
25 MG
$657.00

About This Item

Empirical Formula (Hill Notation):
C19H14N2O4S3
CAS Number:
Molecular Weight:
430.52
UNSPSC Code:
12352200
NACRES:
NA.77

$163.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC(C=C1)=CC=C1SC(OC2=CC=C(N=C(C3=N[C@@H](C(O)=O)CS3)S4)C4=C2)=O

InChI key

XUDRIMTVTPBVLM-CQSZACIVSA-N

Biochem/physiol Actions

Highly sensitive and selective bioluminescent probe suitable for detecting norepinephrine (NE) in vivo
NBP (NE bioluminescent probe) is a highly sensitive and selective bioluminescent probe suitable for detecting norepinephrine (NE) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeping Zuo et al.
Analytical chemistry, 94(17), 6441-6445 (2022-04-23)
As a neurotransmitter, norepinephrine (NE) is critical for psychiatric conditions, neurodegenerative diseases, and pheochromocytoma. A real-time and noninvasive method for the detection of NE as a tracer to investigate the NE-relevant disease treatment process is urgently desirable. Herein, we successfully

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service