Skip to Content
Merck
All Photos(1)

Documents

5.43900

Supelco

Dimethyl sulfoxide

for liquid chromatography LiChrosolv®

Synonym(s):

Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
MDL number:
UNSPSC Code:
51172004
EC Index Number:
200-664-3
NACRES:
NA.03

vapor density

2.7 (vs air)

Quality Level

vapor pressure

0.42 mmHg ( 20 °C)

product line

LiChrosolv®

grade

isocratic

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

HPLC: suitable

impurities

≤0.2% Water

evapn. residue

≤0.002%

transmittance

270 nm, ≥30%
280 nm, ≥60%
300 nm, ≥75%
350 nm, ≥99%

refractive index

n20/D 1.479 (lit.)

kinematic viscosity

2.14 cSt

bp

189 °C (lit.)

mp

16-19 °C (lit.)

transition temp

flash point 87 °C

density

1.10 g/mL (lit.)

storage temp.

2-30°C

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Chemical analysis of acoustically levitated drops by Raman spectroscopy.: Innovative use of acoustical levitation combined with Raman spectroscopy to analyze the chemical properties of droplets in a non-contact setup, useful in scenarios where DMSO is used as a solvent ( Tuckermann et al., 2009).

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Specifications

Saturation Concentration in Air: 8.0 g/m3 (20 °C)

Preparation Note

Product filtered through a 0.5 μm filter

Analysis Note

Purity (GC): ≥ 99.7 %
Water: ≤ 0.2 %
Evaporation residue: ≤ 0.002 %
Transmission (at 270 nm): ≥ 30 %
Transmission (at 280 nm): ≥ 60 %
Transmission (at 300 nm): ≥ 75 %
Transmission (from 350 nm): ≥ 99 %

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service