Skip to Content
Merck
All Photos(1)

Documents

SML3539

Sigma-Aldrich

MS177

≥98% (HPLC)

Synonym(s):

N-((4,6-Dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-6-(6-(4-(2-(3-(2-((2-(2,6- dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4- yl)amino)ethoxy)propanamido)ethyl)piperazin-1-yl)pyridin-3-yl)-1-isopropyl-1H- indazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H55N11O8
CAS Number:
Molecular Weight:
914.02
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(NCC1=C(C=C(NC1=O)C)C)C2=CC(C3=CC=C(N=C3)N4CCN(CC4)CCNC(CCOCCNC5=CC=CC(C(N6C(C(N7)=O)CCC7=O)=O)=C5C6=O)=O)=CC8=C2C=NN8C(C)C

Biochem/physiol Actions

MS177 is a cell penetrant and potent PROTAC degrader of EZH2 (enhancer of zeste homologue 2) that effectively depletes both canonical EZH2–PRC2 and noncanonical EZH2–cMyc complexes. MS177 is fast-acting suppressor of cancer growth. MS177 comprises pomalidomide conjugated to a potent EZH2 inhibitor C24 via a short polyethylene glycol linker. It is much more effective and potent in suppressing cancer growth than catalytic inhibitors of EZH2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EZH2 noncanonically binds cMyc and p300 through a cryptic transactivation domain to mediate gene activation and promote oncogenesis
Nature Cell Biology, 24, 384-399 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service