Skip to Content
Merck
All Photos(1)

Key Documents

SML3096

Sigma-Aldrich

Obeticholic acid

≥98% (HPLC)

Synonym(s):

(3α,5β, 6α,7α)-6-ethyl-3,7-dihydroxy-cholan-24-oic acid, 6-ECDCA, 6a-Ethyl-Chenodeoxycholic Acid, OCA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$161.00
25 MG
$649.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$161.00
25 MG
$649.00

About This Item

Empirical Formula (Hill Notation):
C26H44O4
CAS Number:
Molecular Weight:
420.63
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

$161.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +4 to +6°, c = 1.0 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@@H]([C@@]1([H])CC[C@@]([H])([C@@]2([C@@H]([C@@H]([C@@]3(C[C@@H](CC[C@@]3([C@]2(CC4)[H])C)O)[H])CC)O)[H])[C@@]14C)CCC(O)=O

InChI

1S/C26H44O4/c1-5-17-21-14-16(27)10-12-26(21,4)20-11-13-25(3)18(15(2)6-9-22(28)29)7-8-19(25)23(20)24(17)30/h15-21,23-24,27,30H,5-14H2,1-4H3,(H,28,29)/t15-,16-,17-,18-,19+,20+,21+,23+,24-,25-,26-/m1/s1

InChI key

ZXERDUOLZKYMJM-ZWECCWDJSA-N

Biochem/physiol Actions

Obeticholic acid (6-ECDCA) is a is a semi-synthetic bile acid analogue used to treat used to treat primary biliary cholangitis. Obeticholic acid is an orally available, potent and selective FXR agonist that exhibits anticholeretic and anti-inflammation effects.
orally available, potent and selective FXR agonist that exhibits anticholeretic and anti-inflammation effects

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Pellicciari et al.
Journal of medicinal chemistry, 45(17), 3569-3572 (2002-08-09)
A series of 6alpha-alkyl-substituted analogues of chenodeoxycholic acid (CDCA) were synthesized and evaluated as potential farnesoid X receptor (FXR) ligands. Among them, 6alpha-ethyl-chenodeoxycholic acid (6-ECDCA) was shown to be a very potent and selective FXR agonist (EC(50) = 99 nM)
Botao Gao et al.
Tissue engineering and regenerative medicine, 18(2), 217-224 (2021-02-01)
Ballooned hepatocytes (BH) are a key histological hallmark of nonalcoholic steatohepatitis (NASH), yet their consequences for liver-specific functions are unknown. In our previous study, an experimental model of human induced-BHs (iBH) has been successfully developed based on cell sheet technology. This
Lulu Sun et al.
Nature reviews. Gastroenterology & hepatology, 18(5), 335-347 (2021-02-12)
Farnesoid X receptor (FXR) is a ligand-activated transcription factor involved in the control of bile acid (BA) synthesis and enterohepatic circulation. FXR can influence glucose and lipid homeostasis. Hepatic FXR activation by obeticholic acid is currently used to treat primary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service