Skip to Content
Merck
All Photos(1)

Key Documents

SML3429

Sigma-Aldrich

Maropitant

≥98% (HPLC)

Synonym(s):

(2S,3S)-2-Benzhydryl-N-[(5-tert-butyl-2-methoxyphenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine, (2S,3S)-N-(5-tert-Butyl-2-methoxyphenyl)methyl-2-diphenylmethyl-1-azabicyclo[2.2.2]octan-3-amine, 2-Benzhydryl-1-aza-bicyclo[2.2.2]oct-3-yl)-(5-tert-butyl-2-methoxybenzyl)amine, CJ-11,972, CJ-11972

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00
25 MG
$680.00

$168.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$168.00
25 MG
$680.00

About This Item

Empirical Formula (Hill Notation):
C32H40N2O
CAS Number:
Molecular Weight:
468.67
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$168.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N21[C@H]([C@H](C(CC2)CC1)NCc5c(ccc(c5)C(C)(C)C)OC)C(c4ccccc4)c3ccccc3

InChI

1S/C32H40N2O/c1-32(2,3)27-15-16-28(35-4)26(21-27)22-33-30-25-17-19-34(20-18-25)31(30)29(23-11-7-5-8-12-23)24-13-9-6-10-14-24/h5-16,21,25,29-31,33H,17-20,22H2,1-4H3/t30-,31-/m0/s1

InChI key

OMPCVMLFFSQFIX-CONSDPRKSA-N

Biochem/physiol Actions

Maropitant is a neurokinin-1 receptor (NK-1R) antagonist with in vivo antiemetic efficacy.
Neurokinin-1 receptor (NK-1R) antagonist with in vivo antiemetic efficacy.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service