Skip to Content
Merck
All Photos(1)

Key Documents

SML1720

Sigma-Aldrich

ATI-2341 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

ATI 2341 trifluoroacetate salt, N-(1-Ohexadecyl)-L-methionylglycyl-L-tyrosyl-L-glutaminyl-L-lysyl-L-lysyl-L-leucyl-L-arginyl-L-seryl-L-methionyl-L-threonyl-L-α-aspartyl-L-lysyl-L-tyrosyl-L-arginyl- L-leucine, Pal-MGYQKKLRSMTDKYRL, Pal-Met-Gly-Tyr-Gln-Lys-Lys-Leu-Arg-Ser-Met-Thr-Asp-Lys-Tyr-Arg-Leu

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$418.00

$418.00


Check Cart for Availability


Select a Size

Change View
1 MG
$418.00

About This Item

Empirical Formula (Hill Notation):
C104H178N26O25S2 · xC2HF3O2
CAS Number:
Molecular Weight:
2256.82 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$418.00


Check Cart for Availability

Quality Level

Assay

≥95% (HPLC)

form

film

color

colorless

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

ATI-2341 is a potent and selective allosteric agonist of chemokine CXC-type receptor 4 (CXCR4) that functions as a biased ligand, favoring Gαi activation over Gα13. ATI-2341 does not promote β-arrestin recruitment. ATI-2341 promotes the mobilization of PMNs and HSPCs in the peripheral circulation of both mice and monkeys.
ATI-2341 is a potent and selective allosteric agonist of chemokine CXC-type receptor 4 (CXCR4).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Quoyer et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(52), E5088-E5097 (2013-12-07)
Short lipidated peptide sequences derived from various intracellular loop regions of G protein-coupled receptors (GPCRs) are named pepducins and act as allosteric modulators of a number of GPCRs. Recently, a pepducin selectively targeting the C-X-C chemokine receptor type 4 (CXCR4)
Jesús M Planesas et al.
Journal of molecular graphics & modelling, 60, 1-14 (2015-06-17)
Several examples of allosteric modulators of GPCRs have been reported recently in the literature, but understanding their molecular mechanism presents a new challenge for medicinal chemistry. For the specific case of the cellular receptor CXCR4, it is known that pepducins
Boris Tchernychev et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(51), 22255-22259 (2010-12-09)
The G protein-coupled receptor (GPCR), chemokine CXC-type receptor 4 (CXCR4), and its ligand, CXCL12, mediate the retention of polymorphonuclear neutrophils (PMNs) and hematopoietic stem and progenitor cells (HSPCs) in the bone marrow. Agents that disrupt CXCL12-mediated chemoattraction of CXCR4-expressing cells
Serap Erdogmus et al.
Nature communications, 10(1), 5784-5784 (2019-12-21)
G-protein coupled receptors (GPCRs) are versatile cellular sensors for chemical stimuli, but also serve as mechanosensors involved in various (patho)physiological settings like vascular regulation, cardiac hypertrophy and preeclampsia. However, the molecular mechanisms underlying mechanically induced GPCR activation have remained elusive.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service