Skip to Content
Merck
All Photos(1)

Key Documents

SML3425

Sigma-Aldrich

Harpagoside

≥98% (HPLC)

Synonym(s):

(1S ,4aS ,5R ,7S ,7aS )-1,4a,5,6,7,7a-Hexahydro-4a,5-dihydroxy-7-methyl-7-[[(2E )-1-oxo-3-phenyl-2-propen-1-yl]oxy]cyclopenta[c ]pyran-1-yl β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$523.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$523.00

About This Item

Empirical Formula (Hill Notation):
C24H30O11
CAS Number:
Molecular Weight:
494.49
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -50 to -30°

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2OC=C[C@@]3([C@H]2[C@](C[C@H]3O)(OC(=O)\C=C\c4ccccc4)C)O

InChI

1S/C24H30O11/c1-23(35-16(27)8-7-13-5-3-2-4-6-13)11-15(26)24(31)9-10-32-22(20(23)24)34-21-19(30)18(29)17(28)14(12-25)33-21/h2-10,14-15,17-22,25-26,28-31H,11-12H2,1H3/b8-7+/t14-,15-,17-,18+,19-,20-,21+,22+,23+,24-/m1/s1

InChI key

KVRQGMOSZKPBNS-FMHLWDFHSA-N

Biochem/physiol Actions

Harpagoside is an orally available iridoid glycoside and an active component of Harpagophytum extract with cardioprotective effects. Harpagoside is shown to directly bind to and activate ACE2. Allosterically inhibits ACE2 binding to spike protein and reduces viral entry and lessen SARS-CoV-2 infection. Harpagoside was reported to suppress AP-1, IL-6, COX-1/2, c-FOS, NF-ĸB expression and upregulate BMP2 and Wnt signaling pathways.
Orally available iridoid glycoside that exhibits cardioprotective effects; anti-viral and a direct ACE2 activator

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service