Skip to Content
Merck
All Photos(1)

Key Documents

SML3422

Sigma-Aldrich

Pritelivir

≥98% (HPLC)

Synonym(s):

AIC316, BAY 57-1293, BAY57-1293, N-[5-(Aminosulfonyl)-4-methyl-1,3-thiazol-2-yl]-N-methyl-2-[4-(2-pyridinyl)phenyl]acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$64.10
50 MG
$260.00

$64.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$64.10
50 MG
$260.00

About This Item

Empirical Formula (Hill Notation):
C18H18N4O3S2
CAS Number:
Molecular Weight:
402.49
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$64.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)c1[s]c(nc1C)N(C)C(=O)Cc2ccc(cc2)c3ncccc3

InChI

1S/C18H18N4O3S2/c1-12-17(27(19,24)25)26-18(21-12)22(2)16(23)11-13-6-8-14(9-7-13)15-5-3-4-10-20-15/h3-10H,11H2,1-2H3,(H2,19,24,25)

InChI key

IVZKZONQVYTCKC-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and orally active herpes simplex virus (HSV) helicase-primase inhibitor with antiviral efficacy in cultures and in mice in vivo.
Pritelivir (AIC316; BAY57-1293) is a potent and orally active herpes simplex virus (HSV) helicase-primase inhibitor (IC50 = 30 nM using wt HSV-1F ATPase complex UL5-UL52 & 1 mM ATP) with antiviral efficacy in Vero cultures (replication IC50 = 20 nM against HSV-2G, wt and Acyclovir-resistant HSV-1F; MOI = 0.0025) and in mice in vivo (ED50 = 0.5 mg/kg 3x daily p.o. using HSV-1 Walki or HSV-2 MS), being >28 times more potent than acyclovir and valacyclovir.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service