Skip to Content
Merck
All Photos(1)

Key Documents

475954

Sigma-Aldrich

MRN-ATM Pathway Inhibitor, Mirin

The MRN-ATM Pathway Inhibitor, Mirin, also referenced under CAS 299953-00-7, controls the biological activity of Mre11, component of the MRN (Mre11-Rad50-Nbs1) complex. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

MRN-ATM Pathway Inhibitor, Mirin, (Z)-5-(4-Hydroxybenzylidene)-2-imino-1,3-thiazolidin-4-one, Mirin

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$160.00

$160.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 MG
$160.00

About This Item

Empirical Formula (Hill Notation):
C10H8N2O2S
CAS Number:
Molecular Weight:
220.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$160.00


Estimated to ship on30 May 2025


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red-orange

solubility

DMSO: 10 mg/mL
ethanol: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S1C(=N)NC(=O)C1=Cc2ccc(cc2)O

InChI key

YBHQCJILTOVLHD-UHFFFAOYSA-N

General description

A cell-permeable pyrimidinone compound that inhibits the exonuclease activity of Mre11, component of the MRN (Mre11-Rad50-Nbs1) complex, and prevents MRN-mediated activation of dimeric ATM (IC50 = 12 µM) in cell-free p53 Ser15 phosphorylation assays, while exhibiting little effect against MRN-independent ATM activation in the presence of high DSBs (DNA double strain breaks) or the activity of activated monomeric ATM. Mirin is shown to cause an increase of G2 population in exponentially growing TOSA4 cultures (~1.6-fold at 50 µM) and dramatically increase mitotic cell polulation (~10-fold at 25 µM) after irradiation of G2 phase U2OS cultures by preventing IR-induced G2/M checkpoint. Mirin exhibits no inhibitory activity against Rad50-associated adenylate kinase or Exonulease III.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20&degC.

Other Notes

Garner, K.M., et al. 2009. Nature Chem. Biol.5, 129.
Peterson, S., et al. 2009. Nature Chem. Biol.5, 130.
Dupre, A., et al. 2008. Nature Chem. Biol.4, 119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service