Skip to Content
Merck
All Photos(1)

Key Documents

SML3384

Sigma-Aldrich

Cyclovirobuxine D

≥90% (HPLC)

Synonym(s):

(3α,5α,16α,20S)-4,4,14-Trimethyl-3,20-bis(methylamino)-9,19-cyclopregnan-16-ol, (3β,5α,16α,20S)-4,4,14-trimethyl-3,20-bis(methylamino)-9,19-cyclopregnan-16-ol, Bebuxine, CVB-D

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$131.00
50 MG
$530.00

$131.00


Estimated to ship on08 May 2025



Select a Size

Change View
10 MG
$131.00
50 MG
$530.00

About This Item

Empirical Formula (Hill Notation):
C26H46N2O
CAS Number:
Molecular Weight:
402.66
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$131.00


Estimated to ship on08 May 2025


Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

0.1 M HCl: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

C[C@H](NC)[C@@]1([H])[C@H](O)C[C@@]2(C)[C@]3([H])CC[C@@]4([H])C(C)(C)[C@@H](NC)CC[C@]4(C5)[C@]35CC[C@@]21C

InChI

1S/C26H46N2O/c1-16(27-6)21-17(29)14-24(5)19-9-8-18-22(2,3)20(28-7)10-11-25(18)15-26(19,25)13-12-23(21,24)4/h16-21,27-29H,8-15H2,1-7H3/t16-,17+,18-,19-,20-,21-,23+,24-,25+,26-/m0/s1

InChI key

GMNAPBAUIVITMI-ABNIRSKTSA-N

Biochem/physiol Actions

Cyclovirobuxine D is a steroidal alkaloid component extracted from the root of medical plant Buxus microphylla that exhibits beneficial effect on heart failure, arrhythmias, myocardial ischemia and other cardiovascular diseases. Apparently it increases the release and uptake of Ca(2+) in systolic and diastolic periods. Cyclovirobuxine D induces mitochondria-mediated apoptosis in numerous cancer cells including breast and gastric cancer cells. It induces autophagy-associated cell death via the Akt/mTOR pathway in human breast cancer cells.
Steroidal alkaloid component extracted from the root of medical plant Buxus microphylla that exhibits beneficial effect on cardiovascular diseases

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Yu et al.
Fitoterapia, 83(8), 1653-1665 (2012-10-02)
Our previous studies have shown that cyclovirobuxine D (CVB-D) ameliorated the cardiac function in heart failure rats. Considering the relationship between cardiac function and [Ca(2+)]i, and the role of calcium cycling on regulating [Ca(2+)]i, the present study was designed to
Jie Wu et al.
Molecules (Basel, Switzerland), 20(11), 20659-20668 (2015-11-28)
Gastric cancer is one of the most common malignant cancers, with high death rates, poor prognosis and limited treatment methods. Cyclovirobuxine D (CVB-D) is the main active component of the traditional Chinese medicine Buxus microphylla. In the present study, we
Hanmei Wei et al.
Journal of nanoscience and nanotechnology, 18(8), 5274-5282 (2018-02-21)
The blood-brain barrier (BBB) restricts the delivery of most drugs to the brain. In our previous study, the feasibility of cyclovirobuxine D delivery to the brain by a non-invasive nasal route was evaluated. In this study, a suitable drug delivery

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service