Skip to Content
Merck
All Photos(1)

Key Documents

SML3369

Sigma-Aldrich

A-674563 hydrochloride

≥98% (HPLC)

Synonym(s):

αS-[[[5-(3-methyl-1H-indazol-5-yl)-3-pyridinyl]oxy]methyl]-benzeneethanamine hydrochloride, (1S)-1-Benzyl-2-(5-(3-methyl-1H-indazol-5-yl)-pyridin-3-yloxy)-ethylamine hydrochloride, (2S)-1-(5-(3-methyl-1H-indazol-5-yl)pyridin-3-yloxy)-3-phenylpropan-2-amine hydrochloride, A 674563 hydrochloride, A674563 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$168.00
10 MG
$680.00

$168.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$168.00
10 MG
$680.00

About This Item

Empirical Formula (Hill Notation):
C22H22N4O · xHCl
Molecular Weight:
358.44 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$168.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[nH]1nc(c2c1ccc(c2)c3cncc(c3)OC[C@@H](N)Cc4ccccc4)C

InChI key

BPNUQXPIQBZCMR-IBGZPJMESA-N

Biochem/physiol Actions

A-674563 is an orally available ATP-competitive pan-AKT (PKB, protein Kinase B) inhibitor (AKT1 Ki = 11 nM) with additional potency against PKA, Cdk2 GSK3β (IC50 = 1.4, 4.2,10 nM, respectively). A-674563 reduces the phosphorylation of cellular Akt substrates (0.1-10 μM; MiaPaCa-2) and enhances paclitaxel efficacy in a PC-3 xenograft model in mice in vivo (40 mg A-674563/kg via p.o. b.i.d. for 21 days, 15 mg paclitaxel/day on days 20, 24, 28).
Orally available ATP-competitive pan-AKT (PKB, protein Kinase B) inhibitor that enhances paclitaxel efficacy in a PC-3 xenograft model in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service