Skip to Content
Merck
All Photos(2)

Key Documents

218719

Sigma-Aldrich

Casein Kinase I Inhibitor III, PF-4800567

The Casein Kinase I Inhibitor III, PF-4800567 controls the biological activity of Casein Kinase I. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Casein Kinase I Inhibitor III, PF-4800567, 3-(3-Chlorophenoxymethyl)-1-(tetrahydropyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-ylamine, HCl, CKI Inhibitor III, PF-4800567

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$439.00

$439.00


Estimated to ship on01 June 2025



Select a Size

Change View
5 MG
$439.00

About This Item

Empirical Formula (Hill Notation):
C17H18ClN5O2
CAS Number:
Molecular Weight:
359.81
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

$439.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

NC1=NC=NC2=C1C(COC3=CC=CC(Cl)=C3)=NN2C4CCOCC4

InChI

1S/C17H18ClN5O2/c18-11-2-1-3-13(8-11)25-9-14-15-16(19)20-10-21-17(15)23(22-14)12-4-6-24-7-5-12/h1-3,8,10,12H,4-7,9H2,(H2,19,20,21)

InChI key

AUMDBEHGJRZSOO-UHFFFAOYSA-N

General description

A cell-permeable pyrazolopyrimidinylamino compound that acts as a potent, ATP-competitive and reversible inhibitor of CKIε with ~22-fold greater potency over CKIδ (IC50 = 32 and 711 nM, respectively) and displays excellent selectivity among 50-kinases, with the only exception of EGFR (69% inhibition at 1 µM). Shown to preferentially block PER3 (PERIOD3) nuclear translocation in COS7-GFP-PER3-CKIε cells over CKIδ-transfected cells (IC50 = 0.13 and 2.65 µM, respectively), and PER2 degradation in COS7-GFP-PER2-CKIε cells. Unlike Casein Kinase I Inhibitor II, PF-670462 (Cat. No. 218716), PF-4800567 exerts only marginal effect on circadian clock under similar conditions.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Meng, Q.J., et al. 2010. Proc. Natl. Acad. Sci. USA24, 15240.
Etchegaray, J.P., et al. 2010. PLoS One5, e10303.
Walton, K.M., et al. 2009. J. Pharmacol. Exp. Ther.330, 430.
Badura, L., et al. 2007. J. Pharmacol. Exp. Ther.322, 730.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service