Skip to Content
Merck
All Photos(1)

Key Documents

SML3262

Sigma-Aldrich

TG11-77 Hydrochloride

≥98% (HPLC)

Synonym(s):

2-((4,6-dimethylpyridin-2-yl)amino)- N-(2-(2-methyl-lH-indol-3-yl)ethyl)pyrimidine-5-carboxamide Hydrochloride, 2-[(4,6-dimethyl-2-pyridinyl)amino]-N-[2-(2-methyl-1H-indol-3-yl)ethyl]- 5-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$575.00

$143.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$143.00
25 MG
$575.00

About This Item

Empirical Formula (Hill Notation):
C23H24N6O·HCl
CAS Number:
Molecular Weight:
436.94
UNSPSC Code:
12352200
NACRES:
NA.77

$143.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Brain-permeable, potent and selective EP2 receptor antagonist
TG11-77 is a brain-permeable, water soluble, highly potent, and selective EP2 receptor antagonist. TG11-77 suppress inflammatory gene expression induced by EP2 activation in a microglial cell line. TG11-77 is useful for study CNS diseases in rodent models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potent, selective, water soluble, brain-permeable EP2 receptor antagonist for use in central nervous system disease models.
Radhika Amaradhi, et al.
Journal of Medicinal Chemistry, 63(3), 1032-1050 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service