Skip to Content
Merck
All Photos(1)

Key Documents

506149

Sigma-Aldrich

p53 Activator III, RITA

The p53 Activator III, RITA, also referenced under CAS 213261-59-7, modulates the biological activity of p53. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

p53 Activator III, RITA, NSC 652287, 2,5- bis-(5-Hydroxymethyl-2-thienyl)-furan, Reactivation of p53 and Induction of Tumor cell Apoptosis, MDM2 Antagonist VII, MDM2 Inhibitor X

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$217.00

$217.00


Estimated to ship on24 March 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$217.00

About This Item

Empirical Formula (Hill Notation):
C14H12O3S2
CAS Number:
Molecular Weight:
292.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$217.00


Estimated to ship on24 March 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

ethanol: 1 mg/mL
DMSO: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(ccc1c2[o]c(cc2)c3[s]c(cc3)CO)CO

InChI

1S/C14H12O3S2/c15-7-9-1-5-13(18-9)11-3-4-12(17-11)14-6-2-10(8-16)19-14/h1-6,15-16H,7-8H2

InChI key

KZENBFUSKMWCJF-UHFFFAOYSA-N

General description

A cell-permeable, p53-targeting, tricyclic thiophene derivative that displays antitumor properties. Blocks p53—MDM2 interaction and p53 ubiquitination and induces p53-dependent apoptosis in tumor cells expressing wild-type p53. Shown to restore the tumor suppressor function of p53 by inducing its accumulation in HCT116 colon carcinoma cells (~0.1 µM) and in p53-positive SCID mice xenografted with HCT116 cells (0.1, 1 or 10 mg/kg, ip) in vivo.
A cell-permeable, p53-targeting, tricyclic thiophene derivative that displays antitumor properties. Blocks p53—MDM2 interaction and p53 ubiquitination and induces p53-dependent apoptosis in tumor cells expressing wild-type p53. Shown to restore the tumor suppressor function of p53 by inducing its accumulation in HCT116 colon carcinoma cells (~0.1 µM) and in p53-positive SCID mice xenografted with HCT116 cells (0.1, 1 or 10 mg/kg, ip) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p53
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Grinkevich, V., et al. 2005. Nat. Med.11, 1136.
Krajewski, M., et al. 2005. Nat. Med.11, 1135.
Issaeva, N., et al. 2004. Nat. Med.10, 1321.
Nieves-Neira, W., et al. 1999. Mol. Pharmacol.56, 478.
Rivera, M.I., et al. 1999. Biochem. Pharmacol.57, 1283.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiwei Xie et al.
International journal of biological sciences, 17(9), 2367-2379 (2021-07-10)
Recombinant human bone morphogenetic proteins (rhBMPs) can stimulate bone formation and growth in the treatment of spinal fusions and nonunions. However, it is still unclear whether rhBMPs function in the prevention of intervertebral disc degeneration (IDD). Here, we discovered that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service