Skip to Content
Merck
All Photos(1)

Key Documents

SML3252

Sigma-Aldrich

SF3B1 Protein Degrader

≥95% (HPLC)

Synonym(s):

2-((4-chlorophenyl)amino)-N-(6-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)hexyl)thiazole-4-carboxamide, O4I2-based PROTAC® degrader

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$187.00
25 MG
$756.00

$187.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$187.00
25 MG
$756.00

About This Item

Empirical Formula (Hill Notation):
C29H29ClN6O5S
Molecular Weight:
609.10
UNSPSC Code:
12352202
NACRES:
NA.77

$187.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

under inert gas

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C(C2=C1C=CC=C2NCCCCCCNC(C3=CSC(NC4=CC=C(C=C4)Cl)=N3)=O)=O)C5CCC(NC5=O)=O

InChI key

UWYLVQJEOCYCEW-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable degrader of splicing factor 3B1 (SF3B1)
SF3B1 Protein degrader (reported as "PROTAC-O4I2" in Gama-Brambila et al.) is a cell permeable degrader of splicing factor 3B1 (SF3B1) containing an SF3B1 activator O4I2 connected to the ligand of the cereblon ubiquitin ligase thalidomide. PROTAC-O4I2 selectively degrades SF3B1 in cells. It induces apoptosis of tumor cells and extends survival in Drosophila intestinal tumor model.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo A Gama-Brambila et al.
Cell chemical biology, 28(11), 1616-1627 (2021-05-29)
The proteolysis-targeting chimeras (PROTACs) are a new technology to degrade target proteins. However, their clinical application is limited currently by lack of chemical binders to target proteins. For instance, it is still unknown whether splicing factor 3B subunit 1 (SF3B1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service