Skip to Content
Merck
All Photos(1)

Key Documents

SML3023

Sigma-Aldrich

NR187

≥98% (HPLC)

Synonym(s):

2-((4-Chlorobenzyl)(ethyl)amino)-4-(cyclopentylamino)-N-(3-(2-oxopyrrolidin-1-yl)propyl)pyrimidine-5-carboxamide, NR162 control, NR162 inactive control, NR162 negative control

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$226.00
25 MG
$906.00

$226.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$226.00
25 MG
$906.00

About This Item

Empirical Formula (Hill Notation):
C26H35ClN6O2
Molecular Weight:
499.05
UNSPSC Code:
12352200
NACRES:
NA.77

$226.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

film

color

colorless

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Inactive analog of and negative control for the potent and selective inhibitor NR162 against the calcium/calmodulin-dependent serine protein kinase CASK.
NR187 is an inactive analog of and negative control for the potent and selective inhibitor NR162 against the calcium/calmodulin-dependent serine protein kinase CASK

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service