Skip to Content
Merck
All Photos(1)

Key Documents

SML3222

Sigma-Aldrich

TM-N1324

≥98% (HPLC)

Synonym(s):

4-(2-Chloro-4-fluorophenyl)-1,4,5,7-tetrahydro-3-methyl-1-(9H-purin-6-yl)-6H-pyrazolo[3,4-b]pyridin-6-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$201.00
25 MG
$811.00

$201.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$201.00
25 MG
$811.00

About This Item

Empirical Formula (Hill Notation):
C18H13ClFN7O
CAS Number:
Molecular Weight:
397.79
UNSPSC Code:
12352107
NACRES:
NA.77

$201.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=NN(C(N2)=C1C(C3=C(Cl)C=C(F)C=C3)CC2=O)C4=NC=NC5=C4NC=N5

InChI key

CYQNVXRIWKKWQG-UHFFFAOYSA-N

General description

TM-N1324 also known as Cpd1324[1] is an agonist of GPR39, a G-protein-coupled receptor activated by Zn2+. GPR39 is a member of the ghrelin receptor family that has been implicated in the control of glucose-induced insulin secretion, pancreatic β-cell differentiation, control of lipolysis in white adipose tissue, and may also be involved in the pathophysiology of depression. TM-N1324 is found to activate human GPR39 with potencies of 280 nM and 9 nM in the absence and presence of Zn2+, respectively, and murine GPR39 with potencies of 180 nM and 5 nM in the absence and presence of Zn2+. TM-N1324 is known to stimulate gastric somatostatin secretion, indirectly inhibiting ghrelin secretion. It shows no agonist activity for 156 other GPCRs tested or for related receptors ghrelin and motilin. TM-N1324 serves as a tool to explore the physiological function of GPR39.[1] It stimulates GLP-1 secretion in metabolic studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaare V Grunddal et al.
Molecular metabolism, 49, 101207-101207 (2021-03-13)
Obesity is a complex disease associated with a high risk of comorbidities. Gastric bypass surgery, an invasive procedure with low patient eligibility, is currently the most effective intervention that achieves sustained weight loss. This beneficial effect is attributed to alterations
Thomas M Frimurer et al.
Journal of medicinal chemistry, 60(3), 886-898 (2017-01-04)
The G-protein-coupled receptor 39 (GPR39) is a G-protein-coupled receptor activated by Zn2+. We used a homology model-based approach to identify small-molecule pharmacological tool compounds for the receptor. The method focused on a putative binding site in GPR39 for synthetic ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service