Skip to Content
Merck
All Photos(1)

Key Documents

SML1322

Sigma-Aldrich

Ipamorelin acetate

≥98%

Synonym(s):

2-Methylalanyl-L-histidyl-3-(2-naphthalenyl)-D-alanyl-D-phenylalanyl-L-lysinamide acetate, Aib-His-D-2-Nal-D-Phe-Lys-NH2 acetate, NNC 26-0161 acetate;

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$136.00
50 MG
$438.00

$136.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
$136.00
50 MG
$438.00

About This Item

Empirical Formula (Hill Notation):
C38H49N9O5 · xC2H4O2
CAS Number:
Molecular Weight:
711.85 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

$136.00


Available to ship on17 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

color

white to beige

solubility

H2O: 20 mg/L, clear

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Ipamorelin is a selective growth hormone secretagogue and agonist of the ghrelin receptor that selectively stimulates the ghrelin receptor and causes growth hormone release. The potency and efficacy of ipamorelin are similar to GHRP-6 but without increase plasma levels of ACTH or cortisol. Ipamorelin appears to enter the central nervous system and to activate GRLN receptors in the spinal cord.
Ipamorelin is a selective growth hormone secretagogue and agonist of the ghrelin receptor.
Ipamorelin is known to be useful in reducing the symptoms of postoperative ileus is a gastrointestinal dysfunction arising due to abdominal surgery. Symptoms include abdominal distention, vomiting, nausea, constipation and anorexia.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kalina Venkova et al.
The Journal of pharmacology and experimental therapeutics, 329(3), 1110-1116 (2009-03-18)
Ghrelin and ghrelin mimetics stimulate appetite and enhance gastric motility. The present study investigates whether ipamorelin, a selective growth hormone secretagogue and agonist of the ghrelin receptor, would accelerate gastrointestinal transit and ameliorate the symptoms in a rodent model of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service