Skip to Content
Merck
All Photos(1)

Key Documents

182705

Sigma-Aldrich

AhR Antagonist

The AhR Antagonist, also referenced under CAS 301326-22-7, controls the biological activity of AhR.

Synonym(s):

AhR Antagonist, 2-Methyl-2H-pyrazole-3-carboxylic acid-(2-methyl-4- o-tolyl-azophenyl)-amide, CH-223191, 2-Methyl-2H-pyrazole-3-carboxylic acid-(2-methyl-4-o-tolyl-azophenyl)-amide, CH-223191

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$347.00

$347.00


Estimated to ship on29 May 2025



Select a Size

Change View
10 MG
$347.00

About This Item

Empirical Formula (Hill Notation):
C19H19N5O
CAS Number:
Molecular Weight:
333.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$347.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-brown

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]1(nccc1C(=O)Nc2c(cc(cc2)\N=N\c3c(cccc3)C)C)C

InChI

1S/C19H19N5O/c1-13-6-4-5-7-17(13)23-22-15-8-9-16(14(2)12-15)21-19(25)18-10-11-20-24(18)3/h4-12H,1-3H3,(H,21,25)/b23-22+

InChI key

LKTNEXPODAWWFM-GHVJWSGMSA-N

General description

A cell-permeable pyrazolo-amide compound that potently blocks the binding of TCDD (2,3,7,8-tetrachlorodibenzo-p-dioxin) to AhR (aryl hydrocarbon receptor) in a dose-dependent manner. Shown to Inhibit TCDD-induced AhR transcription activity in HepG2 cells in vitro (IC50 = 30 nM) and prevent TCDD-elicited liver toxicity and wasting syndrome in mice in vivo. Unlike the commonly used AhR antagonist flavone, this inhibitor has no effect on estrogen receptor transcription activity, nor does it exhibit TCDD-like agonist activity even at concentrations as high as 10 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
AhR (aryl hydrocarbon receptor)
Product does not compete with ATP.
Reversible: no
Target IC50: 30 nM in blocking TCDD-induced AhR transcription activity in HepG2 cells in vitro

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Kim, S.H., et al. 2006. Mol. Pharmacol.69, 1871.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service