Skip to Content
Merck
All Photos(1)

Key Documents

SML3123

Sigma-Aldrich

NButGT

≥98% (HPLC)

Synonym(s):

(3aR,5R,6S,7R,7aR)-3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-propyl-5H-pyrano[3,2-d]thiazole-6,7-diol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00
25 MG
$680.00

$168.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$168.00
25 MG
$680.00

About This Item

Empirical Formula (Hill Notation):
C10H17NO4S
CAS Number:
Molecular Weight:
247.31
UNSPSC Code:
41106609
NACRES:
NA.77

$168.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CCCC1=N[C@@H]2[C@H]([C@@H]([C@H](O[C@@H]2S1)CO)O)O

Biochem/physiol Actions

NButGT is a potent and selective inhibitor of O-GlcNAcase (OGA), which elevates O-GlcNAc levels but does not cause insulin resistance in 3T3-L1 adipocytes, and in rats and mice. NButGT does not perturbed glucohomeostasis when used for extended time in rats and mice. NButGT attenuates the accumulation of Aβ, neuroinflammation, and memory in in a mouse model of Alzheimer’s disease. NButGT efficiently increased total O-GlcNAc, improves cardiovascular function and reduced animal mortality in rat model of sepsis.
Specific and potent inhibitor of O-GlcNAcase (OGA)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service